missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human Activin C Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
Brand: Novus Biologicals™ MAB11501-SP
This item is not returnable.
View return policy
Description
Activin C/Inhibin beta C Monoclonal antibody specifically detects Activin C/Inhibin beta C in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry
Specifications
| Activin C/Inhibin beta C | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| inhibin beta C chain, IHBC, INHBC, inhibin, beta C | |
| E. coli-derived recombinant human Activin C, Gly237-Ser352, Accession # P55103 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze/thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Immunohistochemistry, Immunocytochemistry | |
| 1077920 | |
| Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL | |
| P55103 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3626 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction