Learn More
R&D Systems™ Human ADAMTSL2(NP_055509.2) VersaClone cDNA
Specifications
Specifications
| Gene | ADAMTSL2 |
| Accession Number | NP_055509.2 |
| Gene ID (Entrez) | 9719 |
| Gene Alias | ADAMTSL2 ADAMTS-like 2, GPHYSD1 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |