missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Amphiregulin DuoSet ELISA
Description
The Human Amphiregulin DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Amphiregulin. For 15 plates.
Specifications
Specifications
| Quantity | 1 Kit (for 15 Plates) |
| Conjugate | Biotin |
| Assay Range | 15.60 to 1000 pg/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 374 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?