missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human AMPKβ1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
247.00€ - 561.00€
Specifications
| Antigen | AMPK beta 1 |
|---|---|
| Clone | 1092531 |
| Dilution | Western Blot 1 μg/mL, Simple Western 10 μg/mL, Immunohistochemistry 0.5-25 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Simple Western |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30281245
|
R&D Systems
MAB11642-SP |
25 μg |
247.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30281244
|
R&D Systems
MAB11642-100 |
100 μg |
561.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AMPK beta 1 Monoclonal antibody specifically detects AMPK beta 1 in Human samples. It is validated for Western Blot,Immunohistochemistry,Simple WesternSpecifications
| AMPK beta 1 | |
| Western Blot 1 μg/mL, Simple Western 10 μg/mL, Immunohistochemistry 0.5-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 5'-AMP-activated protein kinase beta-1 subunit, AMP-activated protein kinase beta subunit, AMPK, AMPK beta -1 chain, AMPK beta 1,5'-AMP-activated protein kinase subunit beta-1, AMPK subunit beta-1, AMPKb, HAMPKb, MGC17785, PRKAB1, protein kinase, AMP-activated, beta 1 non-catalytic subunit, protein kinase, AMP-activated, noncatalytic, beta-1 | |
| E. coli- derived recombinant human AMPKbeta1, Met1-Ile270, Accession # Q9Y478 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1092531 | |
| Western Blot, Immunohistochemistry, Simple Western | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9Y478 | |
| 5564 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title