missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Angiopoietin-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF623-SP
This item is not returnable.
View return policy
Description
Angiopoietin-2 Polyclonal specifically detects Angiopoietin-2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Angiopoietin-2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AGPT2, ANG2, ANG-2, angiopoietin 2, angiopoietin-2, angiopoietin-2a, angiopoietin-2B, angiopoitin 2, ANGPT2, Tie2-ligand | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 285 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| O15123 | |
| ANGPT2 | |
| Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-2 Asp68-Phe496 Accession # O15123 | |
| 25 μg | |
| Primary | |
| Detects human Angiopoietin-2 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse Angiopoietin-2 is observed,and less than 5% cross-reactivity with recombinant human (rh) Angiopoietin-1 and rhAngiopoietin-4 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction