missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human APH1A Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6908
This item is not returnable.
View return policy
Description
APH1A Monoclonal specifically detects APH1A in Human samples. It is validated for Immunohistochemistry.
Specifications
| APH1A | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| Q96BI3 | |
| APH1A | |
| Human APH1A synthetic peptide SSFERFEIFPKHVTDRSDARLQY Accession # Q96BI3 | |
| 100 μg | |
| Primary | |
| Detects human APH1A in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 665404 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 6530402N02Rik, anterior pharynx defective 1 homolog A (C. elegans), APH-1, APH-1A, aph-1alpha, CGI-78, gamma-secretase subunit APH-1A, Presenilin-stabilization factor, PSF | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 51107 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction