missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BDNF Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC2481V-100UG
This item is not returnable.
View return policy
Description
BDNF Monoclonal specifically detects BDNF in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| BDNF | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P23560 | |
| BDNF | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human BDNF His129-Arg247 Accession # P23560 | |
| RUO | |
| 627 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 35909 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| abrineurin, ANON2, brain-derived neurotrophic factor, BULN2, MGC34632, neurotrophin | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human BDNF in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) NT-3, rhNT-4, or rh beta-NGF is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction