missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Brain Thalamus Whole Tissue Lysate (Adult Whole Multiple Sclerosis)

Product Code. 18227631 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18227631 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18227631 Supplier Novus Biologicals™ Supplier No. NB82059406

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The Human Brain Thalamus Whole Tissue Lysate (Adult Whole Multiple Sclerosis) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot
Tissue Nervous
Quantity 1 mg
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Total Protein - Multiple Sclerosis Disease: Brain: Thalamus
Cellular Fraction Whole
Tissue Condition Multiple Sclerosis
Cross Reactivity Human
Host Species Human
Concentration 5 mg/mL
Product Type Human Brain Thalamus Whole Tissue Lysate (Adult Multiple Sclerosis)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.