missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BTNL8 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB9359-SP
This item is not returnable.
View return policy
Description
BTNL8 Monoclonal specifically detects BTNL8 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry.
Specifications
| BTNL8 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| BTN9.2, butyrophilin-like 8, butyrophilin-like protein 8, UNQ702/PRO1347 | |
| Human embryonic kidney cell line HEK-293-derived Human BTNL8 Gln18-Lys238 Accession # Q6UX41-1 | |
| 25 μg | |
| 79908 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Flow Cytometry | |
| 2187B | |
| Western Blot 2 μg/mL, Simple Western 50 μg/mL, Flow Cytometry 0.25 μg/5 x 10^6 cells | |
| Q6UX41-1 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human BTNL8 in direct ELISAs and Western blots. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction