missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Carbonic Anhydrase IX/CA9 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF2188
This item is not returnable.
View return policy
Description
Carbonic Anhydrase IX/CA9 Polyclonal specifically detects Carbonic Anhydrase IX/CA9 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, CyTOF-reported.
Specifications
| Carbonic Anhydrase IX/CA9 | |
| Polyclonal | |
| Unconjugated | |
| Q16790 | |
| CA9 | |
| Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase IX Pro59-Asp414 Accession # Q16790 | |
| 100 μg | |
| Primary | |
| Detects human Carbonic Anhydrase IX (CA9) in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse CA9 is observed and less than 1% cross-reactivity with recombinant human CA1, 3, 4, 8, 10, 12, and 14 is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 3-15 ug/mL, CyTOF-reported | |
| CA9, CAIX, CA-IX, Carbonate dehydratase IX, carbonic anhydrase 9, carbonic anhydrase IX, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MN, P54/58N, PMW1, RCC, RCC-associated antigen G250, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 768 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto