Learn More
Abcam Human CCL2 (MCP1) knockout HeLa cell line
Code produit 32139492 Tous les produits Abcam Produits
Modifier l'affichage
Click to view available options
Quantity:
2 x 1,000,000 Cells
Conditionnement:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Conditionnement
32139492 2 x 1,000,000 Cells Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 32139492 Fournisseur Abcam Code fournisseur ab2553722x1000000Cells_
 Restricted Item
Estimated Shipment: 07-12-2026
Ajouter au panier
Ajouter au panier
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
CCL2 KO cell line available to order. KO validated by Immunocytochemistry. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp insertion in exon 2 and 5 bp deletion in exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial....
CCL2 KO cell line available to order. KO validated by Immunocytochemistry. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp insertion in exon 2 and 5 bp deletion in exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Spécification
Quantity 2 x 1,000,000 Cells