Learn More
Mouse EPCR Alexa Fluor 488-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems FAB2749G-100UG
This item is not returnable.
View return policy
Description
EPCR Monoclonal specifically detects EPCR in Mouse samples. It is validated for Flow Cytometry.
Specifications
| EPCR | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q64695 | |
| PROCR | |
| Mouse myeloma cell line NS0-derived recombinant mouse EPCR Ala17-Ser214 Accession # Q64695 | |
| RUO | |
| 10544 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 432714 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| APC receptor, CCCA, CCD41, CCD41centrosome-associated protein, CD201, CD201 antigen, centrocyclin, Endothelial cell protein C receptor, endothelial protein C receptor, EPCRMGC23024, PROCR, protein C receptor, endothelial | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse EPCR in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Product Title
For Research Use Only (RUO)