missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NFATC2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6499-SP
This item is not returnable.
View return policy
Description
NFATC2/NFAT1 Monoclonal specifically detects NFATC2/NFAT1 in Human samples. It is validated for Western Blot.
Specifications
| NFATC2/NFAT1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| Q13469 | |
| NFATC2 | |
| E. coli-derived recombinant human NFATC-2 His575-Pro679 Accession # Q13469 | |
| 25 μg | |
| Primary | |
| Detects human NFATC2 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human NFATC1 or C3 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 639402 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| NFAT pre-existing subunit, NFAT transcription complex, preexisting component, NFAT1, NFAT1nuclear factor of activated T-cells, cytoplasmic 2, NFATc2, NF-ATc2, NFATp, NF-ATp, NFATPnuclear factor of activated T-cells, preexisting component, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2, preexisting nuclear factor of activated T-cells 2, T cell transcription factor NFAT1, T-cell transcription factor NFAT1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4773 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction