missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL3/CCL4 Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems IC2701F
This item is not returnable.
View return policy
Description
CCL3/CCL4 Monoclonal specifically detects CCL3/CCL4 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| CCL3/CCL4 | |
| Monoclonal | |
| Fluorescein | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CCL3 | |
| E. coli-derived recombinant human CCL3/MIP-1 alpha Ala27-Ala92 Accession # P10147 | |
| 100 Tests | |
| Primary | |
| Detects human CCL3/MIP-1 alpha in Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) CCL4/MIP-1 beta is observed. Does not cross-react with rhCCL1, 2, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, recombinant mouse CCL1, 2, 3, 5, 6, 7, CCL9/10MIP-1 gamma , 11, 12, 17, 19, 20, 21, 22, 24, 25, or recombinant rat CCL20. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 93342 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| P10147 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6348 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction