missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CD8 beta (NP_757362) VersaClone cDNA

Product Code. 18728119
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18728119 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18728119 Supplier R&D Systems™ Supplier No. RDC1490
  • ONLINE EXCLUSIVE PRICE
    341.00€
    322.35€ / 10µg
    Save 18.65€
    5% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

The Human CD8 beta (NP_757362) VersaClone cDNA clone contains the complete ORF for CD8 beta, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CD8 beta insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature

TRUSTED_SUSTAINABILITY

Specifications

Gene CD8 beta
Accession Number NP_757362
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.163
Insert Type 745bp
Gene ID (Entrez) 926
Gene Alias CD8 antigen, beta polypeptide 1 (p37), CD8 beta, CD8b, CD8b antigen, CD8b molecule, CD8B1, CD8B1P37, LEU2, LY3, LYT3, MGC119115, P37, T lymphocyte surface glycoprotein beta chain, T-cell surface glycoprotein CD8 beta chain
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Product Line R&D Systems
Quantity 10 μg
Show More Show Less

For Reasearch Use Only. Not for use in Humans.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.