missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Chitinase 3-like 1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 7 publications
Brand: R&D Systems AF2599
This item is not returnable.
View return policy
Description
Chitinase 3-like 1 Polyclonal specifically detects Chitinase 3-like 1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| Chitinase 3-like 1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AW208766, Brp39, Chi3l1, Chitinase 3 like 1, gp39, HCgp39, YKL-40 | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| 1116 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.5-1.0 ug/mL, Simple Western 10-25 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 1.5-15 ug/mL | |
| P36222 | |
| CHI3L1 | |
| Mouse myeloma cell line NS0-derived recombinant human Chitinase 3-like 1, Tyr22-Thr383, Accession # P36222 | |
| 100 μg | |
| Primary | |
| Detects human Chitinase 3-like 1 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse Chitinase 3-like 1 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction