missing translation for 'onlineSavingsMsg'
Learn More

Human Chorionic Gonadotropin alpha/beta (HCG) Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

208.00€ - 482.00€

Specifications

Antigen hCG
Clone 1177B
Applications ELISA
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18783357
GREENER_CHOICE
View Documents
R&D Systems
MAB97701-SP
25 μg
208.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18723597
View Documents
R&D Systems
MAB97701-100
100 μg
482.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

hCG Monoclonal specifically detects hCG in Human samples. It is validated for ELISA.
Specifications

Specifications

hCG
ELISA
Unconjugated
Rabbit
Human
P01233 and P01215
IgG
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
1177B
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Chinese Hamster Ovary cell line, CHO-derived human Chorionic Gonadotropin alpha/beta (HCG) and Human CG alpha , Ser21-Gln165 and Ala25-Ser116, Accession # P01233 and P01215
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.