Learn More
R&D Systems™ Human CHRNE (NP_000071) VersaClone cDNA
Product Code. 18770609
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18770609 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18770609 Supplier R&D Systems™ Supplier No. RDC2935
  • ONLINE EXCLUSIVE PRICE
    341.00€
    322.35€ / 10µg
    Save 18.65€
    5% Off
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Contains gene inserts that are fully sequenced and produced with high quality
TRUSTED_SUSTAINABILITY
Specifications
Gene Nicotinic Acetylcholine Receptor epsilon
Accession Number NP_000071
Gene ID (Entrez) 1145
Gene Alias acetylcholine receptor subunit epsilon, AchR epsilon subunit, ACHRE, ACHREFCCMS, cholinergic receptor, nicotinic, epsilon, cholinergic receptor, nicotinic, epsilon polypeptide, CHRNE, CMS1D, CMS1E, CMS2A, nAChR epsilon, SCCMS
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg