missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CILP-1 (AAH35776) VersaClone cDNA
Description
- Also known as: CILP-1; HsT18872
- CILP-1 is a secreted monomeric cartilage glycoprotein.
- CILP-1 is produced by chondrocytes in both hyaline and fibrocartilage, and preferentially found in the inter-territorial matrix of the interior layers of articular cartilage.
- Its expression increases with age and correlates with osteoarthritis.
- The full-length and the Nterminal domain of CILP-1 are shown to function as an IGF-1 antagonist.
- An allelic variant of CILP has been associated with lumbar disc disease.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hCILP |
| Accession Number | AAH35776 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 3568 bp |
| Gene ID (Entrez) | 8483 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?