missing translation for 'onlineSavingsMsg'
Learn More

Human Coronavirus Nucleocapsid Protein Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

183.00€ - 275.00€

Specifications

Antigen Human Coronavirus Nucleocapsid Protein
Dilution Western Blot 1:2000-1:10000, ELISA 1:5000-1:10000
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18649148
View Documents
Novus Biologicals
NBP3-12767-100ul
100 μg
275.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18694259
View Documents
Novus Biologicals
NBP3-12767-50ul
50 μg
183.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Human Coronavirus Nucleocapsid Protein Polyclonal antibody specifically detects Human Coronavirus Nucleocapsid Protein in HCoV-NL63 samples. It is validated for Western Blot, ELISA
Specifications

Specifications

Human Coronavirus Nucleocapsid Protein
Western Blot, ELISA
Unconjugated
Rabbit
HCoV-NL63
Produced in rabbits immunized with purified, recombinant Human Human Coronavirus Nucleocapsid Protein (Accession#: YP_003771.1; Met1-His377)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1:2000-1:10000, ELISA 1:5000-1:10000
Polyclonal
Purified
RUO
0.2 um filtered solution in PBS
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.