Learn More
Human CXCL1/2/3/GRO Pan Specific Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 8 publications
Brand:  R&D Systems MAB276-100
 View more versions of this product
Product Code. 18713187
  • 425.00€ / 100µg
Description
CXCL1/2/3/GRO Monoclonal specifically detects CXCL1/2/3/GRO in Human samples. It is validated for Western Blot, Neutralization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CXCL1/2/3/GRO
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CINC1, CINC-1, CINC-2, CINC3, CINC-3, Dcip1, FSP, Gm1960, GRO alpha, GRO beta, GRO gamma, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, MGSA-beta, MIP-2, NAP-3, SCYB1
E. coli-derived recombinant human CXCL2 Ala35-Asn107 Accession # P19875
100 μg
Primary
Detects human CXCL1/2/3 in direct ELISAs and Western blots.
Human
Ascites
Western Blot, Neutralization
31716
Western Blot 1 ug/mL, Neutralization
P19875
Mouse
Protein A or G purified from ascites
RUO
2919
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.