Learn More
Human CXCR3 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems FAB160G-025
 View more versions of this product
Product Code. 18736129
  • 268.00€ / 25 tests
Description
CXCR3 Monoclonal specifically detects CXCR3 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CXCR3
Monoclonal
Alexa Fluor 488
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXCR3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR
Mouse
Protein A or G purified from ascites
RUO
2833
Human
Purified
Flow Cytometry
49801
Flow Cytometry 5 uL/10^6 cells
P49682
CXCR3
NS0 mouse myeloma cell line transfected with human CXCR3 Met1-Leu368 Accession # P49682
25 Tests
Primary
Detects human CXCR3.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.