missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCR3 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB160G-100
This item is not returnable.
View return policy
Description
CXCR3 Monoclonal specifically detects CXCR3 in Human samples. It is validated for Flow Cytometry.
Specifications
| CXCR3 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXCR3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 2833 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 49801 | |
| Flow Cytometry 5 uL/10^6 cells | |
| P49682 | |
| CXCR3 | |
| NS0 mouse myeloma cell line transfected with human CXCR3 Met1-Leu368 Accession # P49682 | |
| 100 Tests | |
| Primary | |
| Detects human CXCR3. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction