missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCR3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
245.00€ - 507.00€
Specifications
| Antigen | CXCR3 |
|---|---|
| Clone | 49801R |
| Dilution | Flow Cytometry 0.25 μg/10^6 cells, Immunohistochemistry 0.5-25 μg/mL, Neutralization 0.3-1.5 μg/mL, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18190481
![]() |
R&D Systems
MAB160R-SP |
25 μg |
245.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18172041
|
R&D Systems
MAB160R-100 |
100 μg |
507.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCR3 Monoclonal specifically detects CXCR3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-ready.Specifications
| CXCR3 | |
| Flow Cytometry 0.25 μg/10^6 cells, Immunohistochemistry 0.5-25 μg/mL, Neutralization 0.3-1.5 μg/mL, CyTOF-ready | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P49682 | |
| 2833 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 49801R | |
| Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXCR3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR | |
| NS0 mouse myeloma cell line transfected with human CXCR3 Met1-Leu368 Accession # P49682 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CXCR3 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title