missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCR5 PerCP-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
197.00€ - 532.00€
Specifications
| Antigen | CXCR5 |
|---|---|
| Clone | 51505 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18790378
![]() |
R&D Systems
FAB190C-025 |
25 Tests |
197.00€
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18777466
![]() |
R&D Systems
FAB190C-100 |
100 Tests |
532.00€
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCR5 Monoclonal specifically detects CXCR5 in Human samples. It is validated for Flow Cytometry.Specifications
| CXCR5 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| Human | |
| BLR1, BLR1MDR-15, Burkitt lymphoma receptor 1, Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif)receptor 5), Burkitt lymphoma receptor 1, GTP-binding protein, CD185, CD185 antigen, chemokine (C-X-C motif) receptor 5, C-X-C chemokine receptor type 5, CXCR5, CXC-R5, MDR15CXCR-5, MGC117347, Monocyte-derived receptor 15 | |
| CXCR5 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Stains human CXCR5 transfectants but not the parental cell lines in flow cytometry. Does not cross-react with human CXCR2, CXCR3, or CXCR4 transfectants. |
| 51505 | |
| Flow Cytometry | |
| PerCP | |
| Mouse | |
| P32302 | |
| 643 | |
| NS0 mouse myeloma cell line transfected with human CXCR5 Met1-Phe372 Accession # P32302 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title