missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC5L Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-87013IR
This item is not returnable.
View return policy
Description
CDC5L Polyclonal antibody specifically detects CDC5L in Human samples. It is validated for Western Blot
Specifications
| CDC5L | |
| Polyclonal | |
| Western Blot | |
| CDC5, CDC5 (cell division cycle 5, S. pombe, homolog)-like, CDC5 cell division cycle 5-like (S. pombe), Cdc5-like protein, Cdc5-related protein, CEF1, cell division cycle 5-like protein, dJ319D22.1, dJ319D22.1 (CDC5-like protein), hCDC5, KIAA0432, PCDC5RPCDC5-LIKE, Pombe cdc5-related protein | |
| The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human cell division cycle 5 (CDC5) -like using the numbering given in entry NP_001244.1 (GeneID988). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 988 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction