missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DDR2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB25381
This item is not returnable.
View return policy
Description
DDR2 Monoclonal specifically detects DDR2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-reported.
Specifications
| DDR2 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready | |
| Q16832 | |
| DDR2 | |
| Mouse myeloma cell line NS0-derived recombinant human DDR2 Gln24-Arg399 Accession # Q16832 | |
| 100 μg | |
| Primary | |
| Detects human DDR2 in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 290804 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD167 antigen-like family member B, CD167b antigen, Discoidin domain receptor 2, discoidin domain receptor family, member 2, discoidin domain receptor tyrosine kinase 2, discoidin domain-containing receptor 2, EC 2.7.10, EC 2.7.10.1, hydroxyaryl-protein kinase, migration-inducing gene 16 protein, neurotrophic tyrosine kinase receptor related 3, Neurotrophic tyrosine kinase, receptor-related 3, NTRKR3cell migration-inducing protein 20, Receptor protein-tyrosine kinase TKT, TKT, TKTMIG20a, Trk3, Tyro-10, TYRO10, Tyrosine-protein kinase TYRO10, tyrosylprotein kinase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4921 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction