missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Dkk-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
211.00€ - 1825.00€
Specifications
| Antigen | Dkk-1 |
|---|---|
| Clone | 141135 |
| Concentration | LYOPH |
| Dilution | Immunohistochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Immunohistochemistry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18774358
![]() |
R&D Systems
MAB10962-SP |
25 μg |
211.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18711029
![]() |
R&D Systems
MAB10962-100 |
100 μg |
446.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18742819
![]() |
R&D Systems
MAB10962-500 |
500 μg |
1825.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dkk-1 Monoclonal specifically detects Dkk-1 in Human samples. It is validated for Immunohistochemistry, ELISA Capture (Matched Antibody Pair).Specifications
| Dkk-1 | |
| LYOPH | |
| Immunohistochemistry, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| O94907 | |
| 22943 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Dkk-1 Thr32-His266 Accession # O94907 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Dkk-1 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) Dkk-4, rhKremen-1, rhKremen-2 or rhLRP-6 is observed. |
| 141135 | |
| Immunohistochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, Dkk1, DKK-1, hDkk-1, SKdickkopf-1 like | |
| DKK1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title