missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Endoglin/CD105 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 15 publications
Brand: R&D Systems AF1097
This item is not returnable.
View return policy
Description
Endoglin/CD105 Polyclonal specifically detects Endoglin/CD105 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready, Knockout Validated.
Specifications
| Endoglin/CD105 | |
| Polyclonal | |
| Unconjugated | |
| Q5T9B9 | |
| ENG | |
| Mouse myeloma cell line NS0-derived recombinant human Endoglin/CD105 Glu26-Gly586 Accession # Q5T9B9 | |
| 100 μg | |
| Primary | |
| Detects human Endoglin in direct ELISAs and Western blots. In direct ELISAs, less than 15% cross-reactivity with recombinant mouse Endoglin and recombinant rat Endoglin is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF, KnockDown | |
| LYOPH | |
| Western Blot 0.25 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL, CyTOF-ready, Knockout Validated | |
| CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, ENG, HHT1FLJ41744, ORW, ORW1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2022 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction