missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human EphA5 (NP_001268695) VersaClone cDNA
Description
- Also known as: EK7; CEK7; EHK1; HEK7; EHK-1; TYRO4
- EphA5 belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family.
- EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system.
- Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats.
- EphA5 has been shown to bind ephrin-A5, ephrin-A1, ephrin-A2, ephrin-A3, and ephrin-A4.
- The EphA5 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hEPHA5 |
| Accession Number | NP_001268695 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 3064 bp |
| Gene ID (Entrez) | 2044 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?