missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Erythropoietin/EPO Antibody, R&D Systems™

Mouse Monoclonal Antibody
194.00€ - 421.00€
Specifications
| Antigen | Erythropoietin/EPO |
|---|---|
| Clone | 971019 |
| Dilution | Neutralization 0.02-0.2 μg;/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18701457
![]() |
R&D Systems
MAB2872-SP |
25 μg |
194.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18730957
![]() |
R&D Systems
MAB2872-100 |
100 μg |
421.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Human samples. It is validated for Neutralization.Specifications
| Erythropoietin/EPO | |
| Neutralization 0.02-0.2 μg;/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| CAA26094 | |
| 2056.0 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 971019 | |
| Neutralization | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 | |
| Chinese Hamster Ovary cell line CHO-derived human Erythropoietin/EPO protein, Ala28-Arg193, Accession # CAA26094 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Erythropoietin/EPO in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title