missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Erythropoietin R MAb (Clone 713210), R&D Systems™

Mouse Monoclonal Antibody
209.00€ - 425.00€
Specifications
| Antigen | Erythropoietin R |
|---|---|
| Clone | 713210 |
| Dilution | Neutralization 0.4-2.4 ug/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18701418
![]() |
R&D Systems
MAB3073-SP |
25 μg |
209.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18740759
![]() |
R&D Systems
MAB3073-100 |
100 μg |
425.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Erythropoietin R Monoclonal specifically detects Erythropoietin R in Human samples. It is validated for Neutralization.Specifications
| Erythropoietin R | |
| Neutralization 0.4-2.4 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P19235 | |
| 2057 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 713210 | |
| Neutralization | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| EpoR, EPO-R, erythropoietin receptor, MGC138358 | |
| Mouse myeloma cell line NS0-derived recombinant human Erythropoietin R Ala25-Pro250 Accession # P19235 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Erythropoietin R in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title