Learn More
Abcam Human ETS2 knockout HeLa cell line
Artikelnummer. 32155433 Shop alle Abcam produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
1,000,000 Cells
2 x 1,000,000 Cells
Pakningsstørrelse:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Pakningsstørrelse
32155433 1,000,000 Cells Each
32142622 2 x 1,000,000 Cells Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 32155433 Leverandør Abcam Leverandørnr. ab2749451000000Cells_
 Restricted Item
Estimated Shipment: 27-11-2026
Læg i indkøbskurv
Læg i indkøbskurv
missing translation for 'validMainframeMsg'
Denne vare kan ikke returneres. Se returpolicy
TERT KO cell line available to order. KO validated by Next Generation Sequencing. Free of charge wild type control available. 63 bp deletion (allele 1) and 1 bp insertion after Lys168 (allele 2) of the WT protein, Knockout. To order both knockout and wild-type control cells: select 2 x 1000000 C...
TERT KO cell line available to order. KO validated by Next Generation Sequencing. Free of charge wild type control available. 63 bp deletion (allele 1) and 1 bp insertion after Lys168 (allele 2) of the WT protein, Knockout. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Tekniske data
Quantity 1,000,000 Cells
Regulatory Status RUO
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.