missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Exosome Component 9 Antibody, R&D Systems™
Mouse Monoclonal Antibody
234.00€ - 537.00€
Specifications
| Antigen | Exosome Component 9 |
|---|---|
| Clone | 1090209 |
| Dilution | Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279915
|
R&D Systems
MAB11616-SP |
25 μg |
234.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279916
|
R&D Systems
MAB11616-100 |
100 μg |
537.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Exosome Component 9 Monoclonal antibody specifically detects Exosome Component 9 in Human samples. It is validated for Immunohistochemistry,ImmunocytochemistrySpecifications
| Exosome Component 9 | |
| Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| Autoantigen PM/Scl 1, EC 3.1.13, exosome complex component RRP45, exosome complex exonuclease RRP45, P5, p6, P75 polymyositis-scleroderma overlap syndrome associated autoantigen, PCH1D, PM/Scl-75, PMSCL autoantigen, 75kD, PMSCL1, Polymyositis/scleroderma autoantigen 1, polymyositis/scleroderma autoantigen 1 (75kD), polymyositis/scleroderma autoantigen 1, 75kDa, RRP45, Rrp45p | |
| Synthetic Peptide, Accession # Q06265 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1090209 | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q06265 | |
| 5393 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title