missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human G-CSF Antibody, R&D Systems™
Mouse Monoclonal Antibody
232.00€ - 609.00€
Specifications
| Antigen | G-CSF |
|---|---|
| Clone | 741250 |
| Dilution | ELISA, Immunocytochemistry 8 ug/mL |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18119090
|
R&D Systems
MAB2141-SP |
25 μg |
232.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18186131
|
R&D Systems
MAB2141-100 |
100 μg |
609.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
G-CSF Monoclonal antibody specifically detects G-CSF in Human samples. It is validated for ELISA, ImmunocytochemistrySpecifications
| G-CSF | |
| ELISA, Immunocytochemistry 8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3, CSF3OS, Filgrastim, GCSF, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin | |
| E. coli-derived human G-CSF protein, Thr31-Leu204, Accession # P09919 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 741250 | |
| ELISA, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P09919 | |
| 1440 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title