Learn More
Human G-CSFR/CD114 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Marca: R&D Systems AF-381-PB
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
G-CSFR/CD114 Polyclonal specifically detects G-CSFR/CD114 in Human samples. It is validated for Western Blot.
Specifica
| G-CSFR/CD114 | |
| Polyclonal | |
| Unconjugated | |
| Q99062 | |
| CSF3R | |
| Mouse myeloma cell line NS0-derived recombinant human G-CSF R/CD114 Glu25-Pro621 Accession # Q99062 | |
| 100 μg | |
| Primary | |
| Detects human G-CSF R/CD114 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) G-CSF and rhGM-CSF R alpha is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| CD114, CD114 antigen, colony stimulating factor 3 receptor (granulocyte), CSF3R, Csfgr, G-CSF R, G-CSF receptor, GCSFR, GCSFRG-CSF-R, granulocyte colony-stimulating factor receptor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1441 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto