missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPR75 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB67281-100
This item is not returnable.
View return policy
Description
GPR75 Monoclonal antibody specifically detects GPR75 in Human samples. It is validated for Flow Cytometry
Specifications
| GPR75 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| G protein-coupled receptor 75, GPRchr2, MGC132002, probable G-protein coupled receptor 75, WI31133, WI-31133 | |
| NS0 mouse myeloma cell line transfected with human GPR75, Accession # O95800 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |
| Flow Cytometry | |
| 689714 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| O95800 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10936 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction