missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HGF Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
209.00€ - 2080.00€
Specifications
| Antigen | HGF |
|---|---|
| Clone | 24516 |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18746948
![]() |
R&D Systems
MAB694-SP |
25 μg |
209.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18738558
![]() |
R&D Systems
MAB694-100 |
100 μg |
508.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18722049
![]() |
R&D Systems
MAB694-500 |
500 μg |
2080.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HGF Monoclonal specifically detects HGF in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| HGF | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A | |
| HGF | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 24516 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 3082 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human HGF | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects recombinant human (rh)HGF in direct ELISAs and Western blots. In direct ELISAs, less than 0.25% cross-reactivity was observed with rhMSP. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title