Learn More
R&D Systems™ Human HIF-3 alpha/HIF3A (NP_690008.2) VersaClone cDNA
Product Code. 18492350
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18492350 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18492350 Supplier R&D Systems™ Supplier No. RDC3435
Contains gene inserts that are fully sequenced and produced with high quality
TRUSTED_SUSTAINABILITY
Specifications
Gene HIF-3 alpha
Accession Number NP_690008.2
Gene ID (Entrez) 64344
Gene Alias Basic-helix-loop-helix-PAS protein MOP7, BHLHE17, bHLHe17HIF-3A2, Class E basic helix-loop-helix protein 17, HIF3A, HIF-3A4, HIF-3-alpha, HIF3-alpha, hypoxia inducible factor 3, alpha subunit, hypoxia-inducible factor 3-alpha, hypoxia-inducible factor-3 alpha 4, Inhibitory PAS domain protein, IPAS, IPASHIF3-alpha-1, Member of PAS protein 7, MOP7, MOP7PAS domain-containing protein 7, PASD7, PASD7HIF-3A
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg