missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HSP27 (S78/S82) Antibody, R&D Systems™

Rat Monoclonal Antibody
184.00€ - 378.00€
Specifications
| Antigen | HSP27 |
|---|---|
| Clone | 836370 |
| Dilution | Western Blot 0.25 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18707469
![]() |
R&D Systems
MAB23142-SP |
25 μg |
184.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18700569
![]() |
R&D Systems
MAB23142-100 |
100 μg |
378.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSP27 Monoclonal specifically detects HSP27 in Human samples. It is validated for Western Blot.Specifications
| HSP27 | |
| Western Blot 0.25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HSPB1, SRP27, Stress-responsive protein 27 | |
| Phosphopeptide containing human HSP27 S78/S82 sites Accession # P04792 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human HSP27 when dually phosphorylated at S78/S82. |
| 836370 | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Human | |
| P04792 | |
| 3315 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title