missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-alpha/beta R1 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems FAB2452G-100UG
This item is not returnable.
View return policy
Description
IFN-alpha/beta R1 Monoclonal antibody specifically detects IFN-alpha/beta R1 in Human samples. It is validated for Flow Cytometry
Specifications
| IFN-alpha/beta R1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFN-aR1, IFNAR1, IFNBR, IFNbR1, IFN-bR1, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1 | |
| Mouse myeloma cell line, NS0-derived human IFN-alpha/beta R1, Gly26-Lys436, Accession # P17181 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 2951C | |
| Flow Cytometry | |
| P17181 | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3454 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction