missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-gamma Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AB-285-NA
This item is not returnable.
View return policy
Description
IFN-gamma Polyclonal specifically detects IFN-gamma in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| IFN-gamma | |
| Polyclonal | |
| Unconjugated | |
| Q14609 | |
| IFNG | |
| E. coli-derived recombinant human IFN-gamma Met1-Gln144 Accession # Q14609 | |
| 1 mg | |
| Primary | |
| Detects human IFN-gamma in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant rhesus monkey IFN-gamma is observed, and less than 5% cross-reactivity with recombinant mouse IFN-gamma , recombinant canine IFN-gamma , recombinant porcine IFN-gamma , recombinant bovine IFN-gamma , recombinant rat IFN-gamma , recombinant feline IFN-gamma , and recombinant equine IFN-gamma is observed. | |
| Human | |
| Purified |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 1-5 ug/mL | |
| IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| Goat | |
| Protein A or G purified | |
| RUO | |
| 3458 | |
| Reconstitute at 1 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction