missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IgE Antibody (Fc-2001), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-78601
This item is not returnable.
View return policy
Description
Human IgE Monoclonal specifically detects Human IgE in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Radioimmunoassay.
Specifications
| Human IgE | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2b |
| Western Blot, ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
| Fc-2001 | |
| Unconjugated | |
| immunoglobulin heavy constant epsilon | |
| Human Myeloma IgE | |
| 100 μg | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction