missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IgG1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
206.00€ - 493.00€
Specifications
| Antigen | IgG1 |
|---|---|
| Clone | 2347E |
| Dilution | Western Blot 1 μg/mL, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18700217
![]() |
R&D Systems
MAB9930-SP |
25 μg |
206.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18734397
![]() |
R&D Systems
MAB9930-100 |
100 μg |
493.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgG1 Monoclonal specifically detects IgG1 in Human samples. It is validated for Western Blot, ELISA.Specifications
| IgG1 | |
| Western Blot 1 μg/mL, ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| IgG1, Igh-4, RGD1359539, VH7183 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 2347E | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Full length Human IgG1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IgG1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title