missing translation for 'onlineSavingsMsg'
Learn More

Human IgM Antibody (3H10G5F2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

116.00€

Specifications

Antigen Human IgM
Clone 3H10G5F2
Concentration 1 mg/mL
Dilution ELISA 1: 10000
Applications ELISA
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18538582
View Documents
Novus Biologicals
NBP1-51525-0.025mg
0.025 mg
116.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Human IgM Monoclonal specifically detects Human IgM in Human samples. It is validated for ELISA.
Specifications

Specifications

Human IgM
1 mg/mL
ELISA
Unconjugated
RUO
AGM1, MU, VH
IgG1
Affinity Purified
3H10G5F2
ELISA 1: 10000
Monoclonal
Mouse
Human
ChromPure Human IgM (myeloma), whole molecule.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.