Learn More
Human IgM Antibody, R&D Systems™
Rabbit Monoclonal Antibody
209.00€ - 447.00€
Spécification
| Antigen | IgM |
|---|---|
| Clone | 2123C |
| Dilution | Western Blot 0.1 ug/mL, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
18710549
![]() |
R&D Systems
MAB94351-SP |
25 μg |
209.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18746609
![]() |
R&D Systems
MAB94351-100 |
100 μg |
447.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgM Monoclonal specifically detects IgM in Human samples. It is validated for Western Blot, Simple Western, ELISA.Spécification
| IgM | |
| Western Blot 0.1 ug/mL, ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| Immunoglobulin M (IgM), mu chain from human plasma | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IgM in direct ELISAs and Western blots. |
| 2123C | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 3507 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |