missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-12 R beta 2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB19591-SP
This item is not returnable.
View return policy
Description
IL-12 R beta 2 Monoclonal specifically detects IL-12 R beta 2 in Human samples. It is validated for Western Blot.
Specifications
| IL-12 R beta 2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q99665 | |
| IL12RB2 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 2 Lys24-Cys622 Accession # Q99665 | |
| 25 μg | |
| Primary | |
| Detects human IL-12 R beta 2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human IL-12 R beta 1 or recombinant mouse IL-12 R beta 1 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 622509 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL-12 receptor beta 2, IL-12 receptor subunit beta-2, IL12R beta 2, IL-12R subunit beta-2, IL12RB2, IL-12RB2, IL-12R-beta-2, interleukin 12 receptor, beta 2, interleukin-12 receptor beta-2 chain, interleukin-12 receptor subunit beta-2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3595 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction