missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-28 R alpha/IFN-lambda R1 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB5260V-100UG
This item is not returnable.
View return policy
Description
IL-28R alpha/IFN-lambda R1 Monoclonal specifically detects IL-28R alpha/IFN-lambda R1 in Human samples. It is validated for Flow Cytometry.
Specifications
| IL-28R alpha/IFN-lambda R1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8IU57 | |
| IFNLR1 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-28 R alpha/IFN-lambda R1 Arg21-Ala228 Accession # Q8IU57 | |
| RUO | |
| 163702 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 601106 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| class II cytokine receptor CRF2/12, CRF2/12, CRF2-12, Cytokine receptor class-II member 12, Cytokine receptor family 2 member 12, IFN-lambda R1, IFN-lambda receptor 1, IFN-lambda-R1, IFNLR, IFNLR1, IL-28 receptor subunit alpha, IL28R alpha, IL-28R1, IL28RA, IL-28RA, IL-28R-alpha, Interferon lambda receptor 1, interferon lambda, receptor 1, interleukin 28 receptor A, interleukin 28 receptor, alpha, interleukin 28 receptor, alpha (interferon, lambda receptor), interleukin or cytokine receptor 2, interleukin-28 receptor subunit alpha, LICR2, Likely interleukin or cytokine receptor 2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human IL-28 R alpha/IFN-lambda R1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IL-28 alpha is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction