missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTA3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19424
This item is not returnable.
View return policy
Description
MTA3 Polyclonal specifically detects MTA3 in Human samples. It is validated for Western Blot.
Specifications
| MTA3 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal | |
| member 3, metastasis associated 1 family, member 3, metastasis-associated protein MTA3 | |
| Rabbit | |
| 68 kDa | |
| 0.1 mL | |
| Breast Cancer, Cancer | |
| 57504 | |
| Human | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q9BTC8 | |
| MTA3 | |
| Recombinant protein encompassing a sequence within the center region of human MTA3. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction