missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-7R alpha/CD127 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
310.00€ - 552.00€
Specifications
| Antigen | IL-7R alpha/CD127 |
|---|---|
| Clone | 40131 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18701149
![]() |
R&D Systems
FAB306A-025 |
25 Tests |
310.00€
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18726576
![]() |
R&D Systems
FAB306A-100 |
100 Tests |
552.00€
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-7R alpha/CD127 Monoclonal specifically detects IL-7R alpha/CD127 in Human samples. It is validated for Flow Cytometry.Specifications
| IL-7R alpha/CD127 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 3575 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-7 R alpha Extracellular domain | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 40131 | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| Human | |
| CD127, CD127 antigen, CD127ILRA, CDW127, IL-7 R alpha, IL-7 receptor subunit alpha, IL7R, IL7R alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha | |
| IL7R | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Detects human IL-7 R alpha in Western blots. In Western blots, approximately 5% cross-reactivity was observed with recombinant human (rh) IL-10 R, rhIL-2 R beta , rhIL-5 R alpha , and rhIL-6 R. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title